ADVH7F37FA13 AChemBlock

methyl 3-(3,5-dimethylisoxazol-4-yl)propanoate

  • MDL number MFCD08701341
  • Cas Number : 154928-90-2
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-3-5-dimethylisoxazol-4-yl-propanoate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31329
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart