ADVH7F37FA1D AChemBlock

N1-(2-nitrobenzyl)ethane-1,2-diamine dihydrochloride

  • MDL number MFCD22380370
  • Cas Number : 102494-89-3
  • Molecular Weight : 268.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n1-2-nitrobenzyl-ethane-1-2-diamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31340
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart