ADVH7F37FA27 AChemBlock

1-(1,4-dioxaspiro[4.5]decan-8-yl)ethanone

  • MDL number MFCD20257858
  • Cas Number : 35477-39-5
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-1-4-dioxaspiro-4-5-decan-8-yl-ethanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31351
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart