ADVH7F37FA33 AChemBlock

(isoquinolin-6-yl)methanamine hydrochloride

  • MDL number MFCD22392277
  • Cas Number : 1396762-19-8
  • Molecular Weight : 194.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/isoquinolin-6-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31363
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart