ADVH7F37FA34 AChemBlock

4-(2-(4-methoxyphenyl)ethynyl)phenol

  • MDL number MFCD29991226
  • Cas Number : 18938-18-6
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-2-4-methoxyphenyl-ethynyl-phenol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31364
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart