ADVH7F37FA38 AChemBlock

(5-bromo-3-chloropyridin-2-yl)methanamine hydrochloride

  • MDL number MFCD22689891
  • Cas Number : 1416714-02-7
  • Molecular Weight : 257.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-chloropyridin-2-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31368
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart