ADVH7F37FA4E AChemBlock

2-bromo-5-(tributylstannyl)thiazole

  • MDL number MFCD18915051
  • Cas Number : 1447761-46-7
  • Molecular Weight : 453.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-5-tributylstannyl-thiazole.html

Safety Information

signalword Danger
pcodes P301+310;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31391
pictograms GHS06
hstatements H301;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart