ADVH7F37FA4F AChemBlock

methyl 2,3-dihydro-1-hydroxy-1H-indene-5-carboxylate

  • MDL number MFCD13191920
  • Cas Number : 358751-18-5
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-3-dihydro-1-hydroxy-1h-indene-5-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31392
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart