ADVH7F37FA52 AChemBlock

(2-(trifluoromethyl)pyridin-4-yl)methanamine hydrochloride

  • MDL number MFCD12400889
  • Cas Number : 1159813-38-3
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-trifluoromethyl-pyridin-4-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31395
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart