ADVH7F37FA57 AChemBlock

6-bromo-8-methylimidazo[1,2-a]pyrazine

  • MDL number MFCD12033249
  • Cas Number : 1159815-50-5
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-8-methylimidazo-1-2-a-pyrazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31400
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart