ADVH7F37FA5D AChemBlock

3,6-dihydro-2,6-dioxo-2H-pyran-4-yl acetate

  • MDL number MFCD08461730
  • Cas Number : 15997-62-3
  • Molecular Weight : 170.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-6-dihydro-2-6-dioxo-2h-pyran-4-yl-acetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31406
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart