ADVH7F37FA60 AChemBlock

2-(4-nitrophenyl)malononitrile

  • MDL number MFCD00982966
  • Cas Number : 7077-65-8
  • Molecular Weight : 187.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-nitrophenyl-malononitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31409
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart