ADVH7F37FA63 AChemBlock

3,4-dibromo-1-(phenylsulfonyl)-1H-pyrrole

  • MDL number MFCD11501201
  • Cas Number : 1003856-43-6
  • Molecular Weight : 365.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-4-dibromo-1-phenylsulfonyl-1h-pyrrole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31415
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart