ADVH7F37FA64 AChemBlock

2,3-dibromo-4,6-dimethylpyridine

  • MDL number MFCD06253865
  • Cas Number : 610261-10-4
  • Molecular Weight : 264.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/2-3-dibromo-4-6-dimethylpyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=O31416
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart