ADVH7F37FA6A AChemBlock

1-(5-(2,4-difluorophenyl)-2-methyl-1H-pyrrol-3-yl)ethanone

  • MDL number MFCD26607315
  • Cas Number : 1513180-89-6
  • Molecular Weight : 235.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-5-2-4-difluorophenyl-2-methyl-1h-pyrrol-3-yl-ethanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31425
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart