ADVH7F37FA6F AChemBlock

methyl 2-amino-4-(pyridin-2-yl)thiophene-3-carboxylate

  • MDL number MFCD11136556
  • Cas Number : 1019521-30-2
  • Molecular Weight : 234.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-amino-4-pyridin-2-yl-thiophene-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31431
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart