ADVH7F37FA70 AChemBlock

1-(3-fluorophenyl)prop-2-yn-1-ol

  • MDL number MFCD21143266
  • Cas Number : 2107-40-6
  • Molecular Weight : 150.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-fluorophenyl-prop-2-yn-1-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31432
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart