ADVH7F37FA74 AChemBlock

tetrakis(p-tolyloxy)methane

  • MDL number MFCD28656751
  • Cas Number : 54974-05-9
  • Molecular Weight : 440.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tetrakis-p-tolyloxy-methane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31436
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart