ADVH7F37FA7A AChemBlock

(E)-N`-((1H-indol-6-yl)methylene)-2-(2-(tetrahydro-2H-pyran-4-yl)-4,6-dimethylphenoxy)acetohydrazide

  • MDL number MFCD28657178
  • Cas Number : 1572184-68-9
  • Molecular Weight : 405.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/e-n-1h-indol-6-yl-methylene-2-2-tetrahydro-2h-pyran-4-yl-4-6-dimethylphenoxy-acetohydrazide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31442
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart