ADVH7F37FA7B AChemBlock

2,4-dimethoxypyridin-3-amine

  • MDL number MFCD18255875
  • Cas Number : 217096-31-6
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-dimethoxypyridin-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31443
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart