ADVH7F37FA7C AChemBlock

5-methoxy-1,3-dimethyl-1H-benzo[d]imidazol-2(3H)-one

  • MDL number MFCD22549181
  • Cas Number : 64107-38-6
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-1-3-dimethyl-1h-benzo-d-imidazol-2-3h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31444
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart