ADVH7F37FA83 AChemBlock

1-methyl-1H-indazol-6-ol

  • MDL number MFCD00456074
  • Cas Number : 118933-92-9
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1h-indazol-6-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31454
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart