ADVH7F37FA87 AChemBlock

methyl 1,3-dimethylpyrrolidine-3-carboxylate

  • MDL number MFCD26127438
  • Cas Number : 114725-00-7
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-3-dimethylpyrrolidine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31458
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart