ADVH7F37FA89 AChemBlock

8-bromo-6-methyl-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

  • MDL number MFCD23701112
  • Cas Number : 1428532-90-4
  • Molecular Weight : 228.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-6-methyl-1-2-4-triazolo-4-3-a-pyridin-3-2h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31460
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart