ADVH7F37FA93 AChemBlock

1,2-di(pyridin-3-yl)disulfane

  • MDL number MFCD17171436
  • Cas Number : 24367-50-8
  • Molecular Weight : 220.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-2-di-pyridin-3-yl-disulfane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31473
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart