ADVH7F37FAA1 AChemBlock

tert-butyl 3-methyl-5-nitro-1H-indazole-1-carboxylate

  • MDL number MFCD13183004
  • Cas Number : 599183-33-2
  • Molecular Weight : 277.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-methyl-5-nitro-1h-indazole-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31490
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart