ADVH7F37FABB AChemBlock

2-methoxypyridine-3,4-diamine

  • MDL number MFCD10574697
  • Cas Number : 33631-04-8
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methoxypyridine-3-4-diamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31518
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart