ADVH7F37FAC1 AChemBlock

5-bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridin-3-amine

  • MDL number MFCD22689754
  • Cas Number : 1416714-23-2
  • Molecular Weight : 297.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/5-bromo-1-tetrahydro-2h-pyran-2-yl-1h-pyrazolo-3-4-b-pyridin-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=O31524
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart