ADVH7F37FAC6 AChemBlock

2-((pyridin-3-yl)methyl)hydrazine hydrochloride

  • MDL number MFCD11100945
  • Cas Number : 57616-01-0
  • Molecular Weight : 159.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-pyridin-3-yl-methyl-hydrazine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31533
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart