ADVH7F37FACB AChemBlock

1,3-di-tert-butyl-5-heptylbenzene

  • MDL number MFCD28348649
  • Cas Number : 1643540-34-4
  • Molecular Weight : 288.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-di-tert-butyl-5-heptylbenzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31538
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart