ADVH7F37FACE AChemBlock

2-(benzyloxy)-5-bromobenzenamine

  • MDL number MFCD08686860
  • Cas Number : 186797-58-0
  • Molecular Weight : 278.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-benzyloxy-5-bromobenzenamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31541
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart