ADVH7F37FAD3 AChemBlock

3-bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine

  • MDL number MFCD21296836
  • Cas Number : 1021910-58-6
  • Molecular Weight : 251.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-2-2-5-dimethyl-1h-pyrrol-1-yl-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31546
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart