ADVH7F37FAE6 AChemBlock

hex-5-yn-1-amine hydrochloride

  • MDL number MFCD18839147
  • Cas Number : 102169-54-0
  • Molecular Weight : 133.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/hex-5-yn-1-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31570
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart