ADVH7F37FB01 AChemBlock

1-(tetrahydro-2H-pyran-2-yl)-1H-indazol-5-yl-5-boronic acid

  • MDL number MFCD12922966
  • Cas Number : 1562245-02-6
  • Molecular Weight : 246.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-tetrahydro-2h-pyran-2-yl-1h-indazol-5-yl-5-boronic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31604
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart