ADVH7F37FB03 AChemBlock

2-(tributylstannyl)-3-(trifluoromethyl)pyridine

  • MDL number MFCD11109430
  • Cas Number : 1204580-78-8
  • Molecular Weight : 436.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-tributylstannyl-3-trifluoromethyl-pyridine.html

Safety Information

signalword Danger
pcodes P301+310;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31607
pictograms GHS06
hstatements H301;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart