ADVH7F37FB18 AChemBlock

3-(3,5-dimethylisoxazol-4-yl)propanoyl chloride

  • MDL number MFCD15732521
  • Cas Number : 358721-54-7
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-3-5-dimethylisoxazol-4-yl-propanoyl-chloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31632
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart