ADVH7F37FB37 AChemBlock

1-tosyl-1H-indol-7-amine

  • MDL number MFCD22689863
  • Cas Number : 1396762-15-4
  • Molecular Weight : 286.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-tosyl-1h-indol-7-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31668
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart