ADVH7F37FB6F AChemBlock

1,3,5-tri-tert-butyl-2-methoxybenzene

  • MDL number MFCD00182455
  • Cas Number : 3975-80-2
  • Molecular Weight : 276.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-5-tri-tert-butyl-2-methoxybenzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31732
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart