ADVH7F37FB78 AChemBlock

4-chloro-N-cyclopropylpyrimidin-2-amine

  • MDL number MFCD11046853
  • Cas Number : 1044771-76-7
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-n-cyclopropylpyrimidin-2-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31746
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart