ADVH7F37FB79 AChemBlock

2-chloro-N-cyclopropyl-6-methylpyrimidin-4-amine

  • MDL number MFCD16767268
  • Cas Number : 1250038-98-2
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-n-cyclopropyl-6-methylpyrimidin-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31747
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart