ADVH7F37FB7C AChemBlock

(4-methoxyphenyl)-N-((pyridin-2-yl)methyl)methanamine

  • MDL number MFCD01135702
  • Cas Number : 121020-62-0
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-methoxyphenyl-n-pyridin-2-yl-methyl-methanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31751
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart