ADVH7F37FB89 AChemBlock

3-(trifluoromethyl)pyridine-4-carboxamide

  • MDL number MFCD22684694
  • Cas Number : 1416713-20-6
  • Molecular Weight : 190.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-trifluoromethyl-pyridine-4-carboxamide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31768
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart