ADVH7F37FB95 AChemBlock

1-(tetrahydro-2H-pyran-2-yl)-1H-indazole-6-carboxylic acid

  • MDL number MFCD09909303
  • Cas Number : 1017792-97-0
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-tetrahydro-2h-pyran-2-yl-1h-indazole-6-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31781
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart