ADVH7F37FC33 AChemBlock

2-(isopropylamino)acetonitrile hydrochloride

  • MDL number MFCD25509453
  • Cas Number : 38737-90-5
  • Molecular Weight : 134.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-isopropylamino-acetonitrile-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32001
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart