ADVH7F37FC34 AChemBlock

methyl 6-chloro-4-(dimethylamino)pyridine-3-carboxylate

  • MDL number MFCD26744207
  • Cas Number : 1404095-42-6
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-chloro-4-dimethylamino-pyridine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32002
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart