ADVH7F37FC37 AChemBlock

1-(2-methoxy-5-methylphenyl)piperazine hydrochloride

  • MDL number MFCD22570306
  • Cas Number : 189264-72-0
  • Molecular Weight : 242.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-2-methoxy-5-methylphenyl-piperazine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32006
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart