ADVH7F37FC3A AChemBlock

2-(4-chloro-3-fluorophenyl)ethanamine hydrochloride

  • MDL number MFCD29991203
  • Cas Number : 1375067-41-6
  • Molecular Weight : 210.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-chloro-3-fluorophenyl-ethanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32009
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart