ADVH7F37FC3C AChemBlock

4-chloro-5H-pyrimido[4,5-b][1,4]oxazin-6(7H)-one

  • MDL number MFCD27981090
  • Cas Number : 1211524-67-2
  • Molecular Weight : 185.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-5h-pyrimido-4-5-b-1-4-oxazin-6-7h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32011
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart