ADVH7F37FC43 AChemBlock

6-chloro-4-(4-chlorophenyl)pyridine-3-carboxylic acid

  • MDL number MFCD18259171
  • Cas Number : 1256794-74-7
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-4-chlorophenyl-pyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32023
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart