ADVH7F37FC45 AChemBlock

5-methyl-1-(4-methylbenzyl)-1H-1,2,3-triazol-4-amine

  • MDL number MFCD21947922
  • Cas Number : 1498997-64-0
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1-4-methylbenzyl-1h-1-2-3-triazol-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32027
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart